Carbon Planchet Holder, Electron Microscopy Sciences
Carbon Planchet Holder, Electron Microscopy Sciences
Carbon Planchet Holder,1/2"
Catalog # 100492-292
Supplier #:  76284
Carbon Planchet Holder, Electron Microscopy Sciences
Catalog # 100492-292
Carbon Planchet Holder, Electron Microscopy Sciences
Carbon Planchet Holder, Electron Microscopy Sciences
Carbon Planchet Holder,1/2"
Price
$245.18
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Two configurations are offered: 1/2" dia. (12.7 mm) and 1" dia. (25 mm) with 1/8" (3.1 mm) pin.
Specifications
  • Désignation:
    Carbon Planchet Holder,1/2"