Carbon Film on Hexagonal Grids, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
À propos de cet article
A thin film of pure carbon is deposited on the shiny side of the grid.
- Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
- Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together