BEEM® Capsules; Embedding, Size 00, Electron Microscopy Sciences
À propos de cet article
These polyethylene embedding capsules have a 1 x 1 mm face at the tip of a truncated pyramid, each with a hinged cap.
3 Options Available Below
Product Details & Documents
These polyethylene embedding capsules have a 1 x 1 mm face at the tip of a truncated pyramid, each with a hinged cap.
Product Family Options
Informations sur le produit
- DésignationØAvailabilityPrice
- Embedding Capsules Size 008 mmEach$21.59Pack of 100$22.48Supplier Number: 70000-B
- Embedding Capsules Size 008 mmEach$99.28Pack of 500$97.47Supplier Number: 70010-B
- Embedding Capsules Size 008 mmEach$204.41Pack of 1,000$180.34Supplier Number: 70020-B
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Capsule Holder for Size 00
Informations sur le produit
- Catalog #AvailabilityPrice
- Capsule Holder, Clear
Informations sur le produit
- Catalog #AvailabilityPrice
- Capsule Holder for Size 00
Informations sur le produit
- Catalog #AvailabilityPrice
- Embedding Capsule Holder
Informations sur le produit
- Catalog #AvailabilityPrice
- Embedding Capsule Holder
Informations sur le produit
- Catalog #AvailabilityPrice
- Embedding Capsule Holder