Capsule Molds, Electron Microscopy Sciences | Avantor
Capsule Molds, Electron Microscopy Sciences
Capsule Molds, Electron Microscopy Sciences
Certificates
Capsule Molds, Electron Microscopy Sciences
Capsule Molds, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Our molds are made from the strongest possible silicone rubber providing extremely high tear resistance. They are all reusable and produce embedding blocks of numerous shapes and sizes with a wide variety of tips. We also offer a mold which produces blocks to be used for re-embedding.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Capsule Mold Type A, 10 Cavities
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $49.10
    $49.10
    Each
       $49.10
    Supplier Number: 70150
  • Capsule Mold Type C, 12 Cavities
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $70.55
    $70.55
    Each
       $70.55
    Supplier Number: 70161
  • Capsule Mold Type B, 12 Cavities
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $61.08
    $61.08
    Each
       $61.08
    Supplier Number: 70160