Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
À propos de cet article
Made from translucent polypropylene.
- Resistant to breakage
- Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
7 Options Available Below
Product Details & Documents
Made from translucent polypropylene.
- Resistant to breakage
- Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
Product Family Options
Informations sur le produit
- DésignationCapacitéAvailabilityPrice
- Volumetric10 mlEach$364.67Pack of 3$263.16Supplier Number: 70952-10
- Volumetric50 mlEach$596.14Pack of 3$513.31Supplier Number: 70952-50
- Graduated2 mlEach$280.94Pack of 6$219.00Supplier Number: 70951-02
- Volumetric1 mlEach$278.76Pack of 6$239.50Supplier Number: 70952-01
- Volumetric5 mlEach$317.56Pack of 3$242.58Supplier Number: 70952-05
- Volumetric25 mlEach$385.32Pack of 3$294.31Supplier Number: 70952-25
- Volumetric2 mlEach$287.68Pack of 6$219.00Supplier Number: 70952-02