Calibrated Loops, Electron Microscopy Sciences
Calibrated Loops, Electron Microscopy Sciences
Calibrated Loops, Electron Microscopy Sciences
Certificates
Calibrated Loops, Electron Microscopy Sciences
Calibrated Loops, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The 0.01 mL loops are calibrated to A.P.H.A. Standards and are made from 19 gauge B and S platinum wire with 5% rhodium. The 0.001 mL loops is made from 26 gauge B and S platinum with 15% iridium.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Volume
    Longueur
    Availability
    Price
  • Calibrated Loops
    1.45 mm
    0.001 ml
    70 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $463.06
    $463.06
    Each
       $463.06
    Supplier Number: 62430-30
  • Calibrated Loops
    4 mm
    0.01 ml
    25 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $668.11
    $668.11
    Each
       $668.11
    Supplier Number: 62430-10
  • Calibrated Loops
    4 mm
    0.01 ml
    70 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,194.51
    $1,194.51
    Each
       $1,194.51
    Supplier Number: 62430-20