Brushes, Red Sable, Electron Microscopy Sciences
À propos de cet article
Extremely soft and pliable brush is perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
8 Options Available Below
Product Details & Documents
Extremely soft and pliable brush is perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
Product Family Options
Informations sur le produit
- DésignationBristle MaterialTailleAvailabilityPrice
- Supplier Number: 66100-00
- Supplier Number: 66100-01
- Supplier Number: 66100-02
- Supplier Number: 66100-03
- Supplier Number: 66100-30
- Supplier Number: 66100-40
- Supplier Number: 66100-50
- Red Sable Brush Set of 3 Sizes (#2/0; #1 and #2)Sable hair#2/0; #1 and #2Log in pour la disponibilitéEach67,47 $Supplier Number: 66105