Bone Saw, Electron Microscopy Sciences | Avantor
Bone Saw, Electron Microscopy Sciences
Bone Saw
Catalog # 100493-868
Fournisseur #:  72895-20
Bone Saw, Electron Microscopy Sciences
Catalog # 100493-868
Bone Saw, Electron Microscopy Sciences
Bone Saw, Electron Microscopy Sciences
Bone Saw
Price
$427.83
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Bone saw with a rounded end, 203 mm blade.
The inside handle grip is 70 mm and the finger opening is 25 mm.
Spécifications
  • Désignation:
    Bone Saw