Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences | Avantor
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Certificates
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette. The pipettes are graduated for nominal indication of amount of liquid being drawn. Bellows-shaped bulb makes for easy use and provides excellent drop information.
Product Family Options
Informations sur le produit
  • Capacité
    Availability
    Price
  • 60 ml (2 oz.)
    Discontinued
    Supplier Number: 64298-02
  • 250 ml (8.5 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $156.04
    $156.04
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $133.32
    $133.32
    Each
       $156.04
    Pack of 10
       $133.32
    Supplier Number: 64298-08
  • 125 ml (4.2 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $142.07
    $142.07
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $153.61
    $153.61
    Each
       $142.07
    Pack of 10
       $153.61
    Supplier Number: 64298-04