BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
Certificates
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
BEEM™ Embedding Capsules; Special Shapes, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Available in two specially shaped capsules which are designed to overcome commonly encountered embedding problems; such as, small or thin elongated specimens and powder samples. These capsules are also good for centrifuging precipitated matters and allows for less trimming. The bottle neck capsule produces a 7.9 mm OD block with a hemihyperpoloid (rather than faced) tip. The conical capsule produces a 7.9 mm OD block with a conical (rather than faced) tip.
*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Embedding Capsule Bottle Neck Tip
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $23.88
    $23.88
    Log in for availability
    Pack of 100
    Pack of 100 - $24.87
    $24.87
    Each
       $23.88
    Pack of 100
       $24.87
    Supplier Number: 69912-01
  • Embedding Capsule Bottle Neck Tip
    Discontinued
    Supplier Number: 69912-05
  • Embedding Capsule Conical Tip
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $23.06
    $23.06
    Log in for availability
    Pack of 100
    Pack of 100 - $24.00
    $24.00
    Each
       $23.06
    Pack of 100
       $24.00
    Supplier Number: 69913-01
  • Embedding Capsule Conical Tip
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $92.89
    $92.89
    Log in for availability
    Pack of 500
    Pack of 500 - $91.26
    $91.26
    Each
       $92.89
    Pack of 500
       $91.26
    Supplier Number: 69913-05
  • Embedding Capsule Conical Tip
    Discontinued
    Supplier Number: 69913-10
  • Embedding Capsule Bottle Neck Tip
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $161.57
    $161.57
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $165.71
    $165.71
    Each
       $161.57
    Pack of 1,000
       $165.71
    Supplier Number: 69912-10