BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
Certificates
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
BEEM™ Embedding Capsules Size 3 (small), Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These capsules, made from polyethylene, produce blocks with a 5.6 mm O.D. and a pyramid tip. Comes complete with a hinged cap.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Embedding Capsules, Size 3
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $21.82
    $21.82
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $21.96
    $21.96
    Each
       $21.82
    Pack of 100
       $21.96
    Supplier Number: 69910-01
  • Embedding Capsules, Size 3
    Discontinued
    Supplier Number: 69910-05
  • Embedding Capsules, Size 3
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $192.44
    $192.44
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $161.03
    $161.03
    Each
       $192.44
    Pack of 1,000
       $161.03
    Supplier Number: 69910-10
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • BEEM Capsule Holder White for Size 3
    BEEM Capsule Holder White for Size 3
    103301-920
    Log in pour la disponibilité
    Each - $31.32
Informations sur le produit
  • Catalog #
    Availability
    Price
  • BEEM Capsule Holder White for Size 3
    BEEM Capsule Holder White for Size 3
    103302-262
    Log in pour la disponibilité
    Each - $180.15
Informations sur le produit
  • Catalog #
    Availability
    Price
  • BEEM Capsule Holder White for Size 3
    BEEM Capsule Holder White for Size 3
    103305-110
    Log in pour la disponibilité
    Each - $337.62