BEEM™ Capsule Press, Electron Microscopy Sciences | Avantor
BEEM™ Capsule Press, Electron Microscopy Sciences
Capsule Press
Catalog # 100502-164
Fournisseur #:  69920-00
BEEM™ Capsule Press, Electron Microscopy Sciences
Catalog # 100502-164
BEEM™ Capsule Press, Electron Microscopy Sciences
BEEM™ Capsule Press, Electron Microscopy Sciences
Capsule Press
Price
$1,071.60
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Capsule Press is designed to facilitate the removal of blocks from the capsule without any damage to the block tip.
Ordering information: For BEEM®* capsule size #3, an adapter is required.
Spécifications
  • Désignation:
    Capsule Press