BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
Certificates
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
BEEM™ Dial-A-Grid Storage Modules, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A two tone color coded plastic box with insert which has 24 letter-labeled crossed slots, where the grids can be stored. A rotating protection plate covers the slots and allows for exposure of one grid at a time. Measures L x W x H: 57 x 45 x 12.5 mm (2 1/3 x 1 3/4 x 1/2").
*BEEM® Is A Registered Trademark of Better Equipment For Electron Microscopy, Inc.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Dial-A-Grid Box
    Discontinued
    Supplier Number: 71148-10
  • Dial-A-Grid Box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 813,41 $
    813,41 $
    Log in pour la disponibilité
    Pack of 50
    Pack of 50 - 737,99 $
    737,99 $
    Each
       813,41 $
    Pack of 50
       737,99 $
    Supplier Number: 71148-05
  • Dial-A-Grid Box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 19,53 $
    19,53 $
    Each
       19,53 $
    Supplier Number: 71148-01