Angle Shield, Electron Microscopy Sciences
Product Details & Documents
Splash protecting in standing or sitting position provides good protection personal from pipetting or pouring chemicals.
- Acrylic base work area is 50.8×30.5 cm (20×12") deep
- Face is 50.9×38 cm (20×15") high
An extra wide, angled viewing area and low bend allow the shield to be used from a standing or sitting position. Made from 6.4 mm (¼") thick clear acrylic with non-skid feet for stability.
: Not recommended for Beta use.
Specifications
- Désignation:Angle Shield (New Style)