LEO SEM Mount, Electron Microscopy Sciences
LEO SEM Mount, Electron Microscopy Sciences
LEO SEM Mount, Electron Microscopy Sciences
LEO SEM Mount, Electron Microscopy Sciences
These mounts with tapered end pin
  • Slotted head 12.7 mm (1/2") diameter
  • Pin 3.1 x 5 mm (1/8 x 1/4")
Product Family Options
  • Désignation
    Taille
  • LEO Microscope Mounts Slotted Head
    50
    Each
    Each - $84.02
    $84.02
    Pack of 50
    Pack of 50 - $70.61
    $70.61
    Each
       $84.02
    Pack of 50
       $70.61
     : 75190
  • LEO Microscope Mounts Slotted Head
    100
    Each
    Each - $172.94
    $172.94
    Pack of 100
    Pack of 100 - $144.73
    $144.73
    Each
       $172.94
    Pack of 100
       $144.73
     : 75191