Absorbant Paper Points, Electron Microscopy Sciences
Absorbant Paper Points, Electron Microscopy Sciences
Absorbant Paper Points, Electron Microscopy Sciences
Certificates
Absorbant Paper Points, Electron Microscopy Sciences
Absorbant Paper Points, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Machine rolled paper with a tapered end which offers good water absorbency, and assists you in the removal of surplus water from the knife boat, on the block face, and from the grids. Available in two assortment sizes. The bigger the number the larger the point.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Paper Points, Selection 1
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 200
    Box of 200 - $70.12
    $70.12
    Log in for availability
    Each
    Each - $70.12
    $70.12
    Box of 200
       $70.12
    Each
       $70.12
    Supplier Number: 71011-01
  • Paper Points, Selection 2
    Log in for availability
    Log in for availability
    Log in for availability
    Box of 200
    Box of 200 - $58.31
    $58.31
    Log in for availability
    Each
    Each - $58.31
    $58.31
    Box of 200
       $58.31
    Each
       $58.31
    Supplier Number: 71011-02