Ampoule Cracker, Electron Microscopy Sciences
Ampoule Cracker, Electron Microscopy Sciences
Certificates
Ampoule Cracker, Electron Microscopy Sciences
Ampoule Cracker, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Ampoule Cracker™ is a disposable plastic fracture and safety collar, which is specially designed to aid in the breaking open of pre-scored glass ampoules.
Simply slip the Ampoule Cracker™ over the top at the pre-scored portion, snap it, and discard both the ampoule top and the safety collar to prevent cross contamination.
Product Family Options
Informations sur le produit
  • Désignation
    For
    Availability
    Price
  • Ampoule Breaker
    1 - 4 ml ampoules
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $138.58
    $138.58
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $122.36
    $122.36
    Each
       $138.58
    Pack of 500
       $122.36
    Supplier Number: 60605-04
  • Ampoule Breaker
    5 - 10 ml ampoules
    Discontinued
    Supplier Number: 60605-10
  • Ampoule Breaker
    10 - 20 ml ampoules
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $301.16
    $301.16
    Log in pour la disponibilité
    Pack of 500
    Pack of 500 - $223.32
    $223.32
    Each
       $301.16
    Pack of 500
       $223.32
    Supplier Number: 60605-20