Microscope Slide Mailer for 5 Slides, CMS Labcraft, Electron Microscopy Sciences
À propos de cet article
Holds up to 5 slides 3x1"in size . These mailers are rigid, strong and durable with a convenient flip-top, snap-on lid.
When it stands in its upright position, it can be used as a staining container, with a minimal amount of solution being used. Packaged 25 mailers/pk, 8 packs/cs.
4 Options Available Below
Product Details & Documents
Holds up to 5 slides 3x1"in size . These mailers are rigid, strong and durable with a convenient flip-top, snap-on lid.
When it stands in its upright position, it can be used as a staining container, with a minimal amount of solution being used. Packaged 25 mailers/pk, 8 packs/cs.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Model 1Each$332.20Pack of 200$246.97Supplier Number: 71548-08
- Model 2Each$346.18Pack of 200$262.11Supplier Number: 71549-08
- Model 2Each$40.14Pack of 25$36.40Supplier Number: 71549-01
- Model 1Each$41.34Pack of 25$35.01Supplier Number: 71548-01