Microscope Slide Container for 5 Slides, Electron Microscopy Sciences
Microscope Slide Container for 5 Slides, Electron Microscopy Sciences
Microscope Slide Container
Catalog # 100488-442
Supplier #:  71550
Microscope Slide Container for 5 Slides, Electron Microscopy Sciences
Catalog # 100488-442
Microscope Slide Container for 5 Slides, Electron Microscopy Sciences
Microscope Slide Container for 5 Slides, Electron Microscopy Sciences
Microscope Slide Container
Price
$53.86
Log in for availability
Each Pack of 25
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The microscope slide container holds 5 slides without contact with one another. Ideal for air conveyor transport system. Measures: 3 L x 5/8 W x 11/4" H (51 x 16 x 32 mm).
Specifications
  • Désignation:
    Microscope Slide Container