Five-Channel Timer, Electron Microscopy Sciences | Avantor
Five-Channel Timer, Electron Microscopy Sciences
Five-Channel Timer, Electron Microscopy Sciences
Certificates
Five-Channel Timer, Electron Microscopy Sciences
Five-Channel Timer, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours.
  • Memory recalls last timer setting for reuse
  • Has a clock mode
Its extra large display indicates channels that are in use. Each one has a distinctive 1 minute alarm. Automatically begins to count up to indicate time elapsed after count down reaches zero.
Dimensions: 2⁷/₈×2¹/₂×⁹/₁₆"
Weight: 2 oz.
 : Comes complete with a stand, pocket clip, and magnet.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Timer 5-channel certified
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $206.46
    $206.46
    Each
       $206.46
    Supplier Number: 72600-02
  • Timer, 5-channel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $107.11
    $107.11
    Each
       $107.11
    Supplier Number: 72600