Secured Hinge 100-Capacity Microscope Slide Boxes, Electron Microscopy Sciences
À propos de cet article
Made from durable polystyrene (with heavy walls).
This material is unaffected by humidity and is thoroughly insect proof. Each box is secured with a rust-resistant nickel-plated clasp and hinge pin. The cork lining prevents damage to the stored slides. To ensure proper slide identification, each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. To save space the boxes are stackable. Each box stores up to 100 3x1" or 25 x 75 mm microscope slides. available in 5 color coded boxes making the boxes easy to identify. Each box measures 222 x 172 x 35 mm (8.75 x 6.75 x 3.375").
10 Options Available Below
Product Details & Documents
Made from durable polystyrene (with heavy walls).
This material is unaffected by humidity and is thoroughly insect proof. Each box is secured with a rust-resistant nickel-plated clasp and hinge pin. The cork lining prevents damage to the stored slides. To ensure proper slide identification, each slot is numbered to correspond to the slide inventory sheet on the inside cover of the box. To save space the boxes are stackable. Each box stores up to 100 3x1" or 25 x 75 mm microscope slides. available in 5 color coded boxes making the boxes easy to identify. Each box measures 222 x 172 x 35 mm (8.75 x 6.75 x 3.375").
Product Family Options
Informations sur le produit
- DésignationColorAvailabilityPrice
- 100 CapacityYellowEach$257.81Pack of 10$197.18Supplier Number: 71471-Y
- 100 CapacityGreenEach$259.04Pack of 10$198.12Supplier Number: 71471-G
- 100 CapacityRedEach$218.39Pack of 10$198.12Supplier Number: 71471-R
- Supplier Number: 71470-G
- Supplier Number: 71470-W
- Supplier Number: 71470-Y
- 100 CapacityDiscontinued-Supplier Number: -
- Supplier Number: 71470-B
- Supplier Number: 71470-R
- 100 CapacityBlueEach$259.04Pack of 10$198.12Supplier Number: 71471-B
- 100 CapacityWhiteEach$261.94Pack of 10$209.32Supplier Number: 71471-W