Microscope Slide Box for 100 Slides, Electron Microscopy Sciences
Microscope Slide Box for 100 Slides, Electron Microscopy Sciences
Microscope Slide Box for 100 Slides, Electron Microscopy Sciences
Certificates
Microscope Slide Box for 100 Slides, Electron Microscopy Sciences
Microscope Slide Box for 100 Slides, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PLEASE NOTE: For file up to 1,200 1x3" slides, refer to .
Molded one-piece polypropylene construction with an integral living hinge. Numbering in the base corresponds to the printed index in the covers for slide identification purpose. Boxes are designed to be stackable to save space. The tensioned clasp insures a secure closure. Available in 25 or 100 capacity. Fits 1 x 3" microscope slides.
PLEASE NOTE: For file up to 1,200 1x3" slides, refer to .
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Molded, 100- Microscope Slide Box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $24.72
    $24.72
    Each
       $24.72
    Supplier Number: 71472
  • Molded, 100- Microscope Slide Box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $236.20
    $236.20
    Each
       $236.20
    Supplier Number: 71473