ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
Certificates
ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
8 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Overall Length
    Tip Style
    Availability
    Price
  • Style 3C SA/Dr
    SA steel
    110 mm
    Very sharp and superior finish
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $108.66
    $108.66
    Each
       $108.66
    Supplier Number: 78393-3C
  • Style 5 SA/Dr
    SA steel
    110 mm
    Very sharp and superior finish
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $119.37
    $119.37
    Each
       $119.37
    Supplier Number: 78395-5
  • Style 2A SA/Dr
    SA steel
    115 mm
    Flat round tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $119.45
    $119.45
    Each
       $119.45
    Supplier Number: 78392-2A
  • Style 7 SA/Dr
    SA steel
    115 mm
    Fine and curve tips
    Discontinued
    Supplier Number: 78397-7
  • Style 00 SA/Dr
    SA steel
    120 mm
    Flat edges and strong tips
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $105.69
    $105.69
    Each
       $105.69
    Supplier Number: 78390-00
  • Style 3 SA/Dr
    SA steel
    120 mm
    Very sharp and superior finish
    Discontinued
    Supplier Number: 78393-3
  • Style 4 rubber coated
    Stainless steel, anti-magnetic, anti-acid
    110 mm
    Straight, extra fine
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $145.69
    $145.69
    Each
       $145.69
    Supplier Number: 78325-4DR
  • Style 51S rubber coated
    Stainless steel, anti-magnetic, anti-acid
    115 mm
    Bent for maximum visibility extra fine
    Discontinued
    Supplier Number: 78325-16
    Spécifications:
    Marketsource
    Marketsource
  • Style 5A rubber coated
    Stainless steel, anti-magnetic, anti-acid
    115 mm
    Straight, extra fine
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $148.65
    $148.65
    Each
       $148.65
    Supplier Number: 78325-5A
  • Style 5C rubber coated
    Stainless steel, anti-magnetic, anti-acid
    115 mm
    Extra fine, double bent
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $151.81
    $151.81
    Each
       $151.81
    Supplier Number: 78325-5C
  • Style 2AB rubber coated
    Stainless steel, anti-magnetic, anti-acid
    120 mm
    Flat, round, curved, superior finish
    Discontinued
    Supplier Number: 78325-14
    Spécifications:
    Marketsource
    Marketsource
  • Style AA rubber coated
    Stainless steel, anti-magnetic, anti-acid
    130 mm
    Straight, sharp, fine
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $107.46
    $107.46
    Each
       $107.46
    Supplier Number: 78325-AA