EMS Flat-Tip Tweezers, Electron Microscopy Sciences
À propos de cet article
These long and slender steel tweezers work well in multiple precision applications and are especially well suited for working near heat sources.
- Made from SA steel
- Serrated grips
10 Options Available Below
Product Details & Documents
These long and slender steel tweezers work well in multiple precision applications and are especially well suited for working near heat sources.
- Made from SA steel
- Serrated grips
Product Family Options
Informations sur le produit
- DésignationLongueurTip StyleAvailabilityPrice
- Style 34A120 mmSmooth and wide tips, good for handling delicate partsLog in pour la disponibilitéEach$70.30Supplier Number: 78334-34A
- Supplier Number: 78211-01
- Style 35B118 mmSmooth and short curved wide tipsEach$590.52Pack of 6$464.44Supplier Number: 78211-06
- Supplier Number: 78333-33A
- Straight Spade Tip Forcepts, Type 127105 mmThin and square tips, straightLog in pour la disponibilitéEach$73.62Supplier Number: 72972-SP
- Supplier Number: 78332-125
- Style 125A120 mmSmooth, flat, and elongated, no scratch tipsLog in pour la disponibilitéEach$66.39Supplier Number: 78332-125A
- Supplier Number: 78335-35A
- Angle Spade Tip Forceps, Type 128105 mmThin and square tips, straightLog in pour la disponibilitéEach$79.94Supplier Number: 72972-AP
- Style 36A120 mmSmooth and upward bent wide tips, good for handling delicate partsLog in pour la disponibilitéEach$67.37Supplier Number: 78336-36A
- Style 574A145 mmFlat and tips bent upwardDiscontinued-Supplier Number: 78212-06