EMS Surface Mount and Optoelectric Tweezers, Electron Microscopy Sciences
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements.
They offer comfort and smooth handling.They are available in different tips and angles for specific jobs and they have a satin antiglare finish with an ergonomic design.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together