Microcentrifuge Polyallomer Tube with Snap-On Cap, Beckman Coulter
Catalogue # CABK357448
Fournisseur: Beckman Coulter
Spécifications
- Volume:1.5 ml
- Description:Polypropylene Tube
- Dimensions:11 x 38 mm
- Cat. no.:CABK357448
- Supplier No.:357448
Spécifications
À propos de cet article
Microfuge tubes are required for use in the Biomek® 2000/3000 systems.
- Compatible with inserts
- For Biomek 2000/3000
Specially designed to be compatible with 11 mm inserts, these tubes with inserts fit into the 24-place Rack for use on the Biomek.