CaptairBio PCR Workstations, Erlab
CaptairBio PCR Workstations, Erlab
   Options durables disponibles
Supplier:   Erlab
CaptairBio PCR Workstations, Erlab
Supplier:  Erlab
   Options durables disponibles
Certificates
CaptairBio PCR Workstations, Erlab
4 Options Available Below
Product Details & Documents
Erlab’s CaptairBio PCR Workstations are designed to protect sensitive biological applications against both environmental pollution and cross-contamination. The HEPA filter will bring clean air down to the work surface, with high efficiency UV lights to decontaminate the worktop from biological cross-contamination.
  • Each unit has Smart Technology™ - an exclusive set of tools that consists of Smart-Light Communication™, chemical sensors (*optional if adding carbon filters), real-time status, and the eGuard app
  • Smart Technology gives an easily identifiable method of communication regarding the sash being opened while the UV light is on, air face velocity is compromised: check sash, pre-filter, or HEPA/ULPA, and fan status via a soft, LED band of light called Smart-Light
  • Smart-Light is a light signature across the front panel which casts a stable LED glow ensuring proper operation
If normal operation is disrupted, the reassuring LED light pulses and sounds, drawing the attention of the operator only when necessary for the following:
2 beeps- Sash open with UV light on
3 beeps- Air Face Velocity is compromised; check sash, pre-filter, or HEPA/ULPA filter
4 beeps- Fan Failure
AirProtect 320 PCR Workstation will feature internal LED and UV lights only, the system will not feature a HEPA filter or will require replacement HEPA filters. The stainless steel work surface cost is not included, VWR# 10170-026.
Certifications: CE, ISO 5 (when used in a cleanroom in compliance with ISO 9/EN 14-644 standard).
Ordering information: All models require HEPA filters in order to operate with the ability to add carbon filters if necessary. Filters and accessories are sold separately. Units are supplied with a manufacturer's lifetime warranty (subject to compliance with operating conditions). The units will arrive completely knocked down and will require assembly. Please contact your Avantor representative for more information.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 52428-824
  • 52428-826
  • 10170-026
  • 10170-030
  • Environmentally Preferable:
  • Energy Efficient
  • Energy Efficient
  • Airflow:
  • 183 cfm
  • 183 cfm
  • Electrical:
  • 115 V/60 Hz or 230/50 Hz
  • 115 V/60 Hz or 230/50 Hz
  • Structure:
  • Anti-Corrosion Steel with 100% Polyester; Acrylic Panels; Injected Polypropylene Filtration
  • Anti-Corrosion Steel with 100% Polyester; Acrylic Panels; Injected Polypropylene Filtration
Spécifications
Product Family Options
Workstations
Informations sur le produit
  • Désignation
    Inside Depth
    Inside Height
    Inside Width
    Outside Height
    Outside Width
    Availability
    Price
  • CaptairBio 321 PCR workstation with 1 bactericidal UV lamp, 15 W
    24¾"
    20¹¹/₁₆"
    30⅛"
    34⁷/₈" min., 38" max.
    32¼"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,442.38
    $7,442.38
    Each
       $7,442.38
    Minimum order quantity is 1
    Supplier Number: 321PCR
    Spécifications:
    Small Business Enterprise
      Energy Efficient
    More
    Specifications
    Cat. No.
    52428-824
    Environmentally Preferable
    Energy Efficient
    Airflow
    183 cfm
    Electrical
    115 V/60 Hz or 230/50 Hz
    Structure
    Anti-Corrosion Steel with 100% Polyester; Acrylic Panels; Injected Polypropylene Filtration
    Small Business Enterprise
      Energy Efficient
  • CaptairBio 391 PCR workstation with 2 bactericidal UV lamps, 15 W each
    24¾"
    23⁷/₁₆"
    38³⁄₁₆"
    37¼" min., 40³/₈" max.
    40⁹/₁₆"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,938.64
    $8,938.64
    Each
       $8,938.64
    Minimum order quantity is 1
    Supplier Number: 391PCR
    Spécifications:
    Small Business Enterprise
      Energy Efficient
    More
    Specifications
    Cat. No.
    52428-826
    Environmentally Preferable
    Energy Efficient
    Airflow
    183 cfm
    Electrical
    115 V/60 Hz or 230/50 Hz
    Structure
    Anti-Corrosion Steel with 100% Polyester; Acrylic Panels; Injected Polypropylene Filtration
    Small Business Enterprise
      Energy Efficient
Components
Informations sur le produit
  • Désignation
    Inside Depth
    Inside Height
    Inside Width
    Outside Height
    Outside Width
    Availability
    Price
  • Bio 321 Stainless Steel Worksurface
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,029.39
    $1,029.39
    Each
       $1,029.39
    Minimum order quantity is 1
    Supplier Number: 321SST
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    10170-026
    Environmentally Preferable
    Airflow
    Electrical
    Structure
    Small Business Enterprise
  • Bio 391 Stainless Steel Worksurface
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,251.47
    $1,251.47
    Each
       $1,251.47
    Minimum order quantity is 1
    Supplier Number: 391SST
    Spécifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    10170-030
    Environmentally Preferable
    Airflow
    Electrical
    Structure
    Small Business Enterprise
Filters
Informations sur le produit
  • Catalog #
    For Use With
    Désignation
    Availability
    Price
  • Captair Smart, Flow, Bio, Flex M and XLS, Store 834 and 1634
    BE+: Carbon filter for use with Acids, Organics, In-organics and bases.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,371.43
    $1,371.43
    Each
       $1,371.43
Informations sur le produit
  • Catalog #
    For Use With
    Désignation
    Availability
    Price
  • Captair Smart, Flow, Bio, Flex M and XLS, Store 834 and 1634
    AS: Carbon filter for use with volatile organic compounds
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $935.36
    $935.36
    Each
       $935.36
Informations sur le produit
  • Catalog #
    For Use With
    Désignation
    Availability
    Price
  • Captair Smart, Captair Bio, Captair Flow 321 and 391 (Under Fan Box), Captair Flow 321/391/483/714 (Over Fan Box), Captair Flex S and M, Captair Store 834 and 1634
    Prefilter M - Protects Hepa and carbon filters from dust contained in the laboratory environment
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $84.30
    $84.30
    Each
       $84.30
Informations sur le produit
  • Catalog #
    For Use With
    Désignation
    Availability
    Price
  • Captair Smart, Flow, Bio, Flex M and XLS, Store 834 and 1634
    HP: Hepa H14 filter provides 99.995% efficiency for trapping 0.1 Micron particles or higher
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $992.10
    $992.10
    Each
       $992.10