Axygen® 24-Well Deep Well Plates, Rectangular Wells, Corning | Avantor
Axygen® 24-Well Deep Well Plates, Rectangular Wells, Corning
   Options durables disponibles
Supplier:   Corning
Axygen® 24-Well Deep Well Plates, Rectangular Wells, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
Axygen® 24-Well Deep Well Plates, Rectangular Wells, Corning
Axygen® 24-Well Deep Well Plates, Rectangular Wells, Corning
2 Options Available Below
Product Details & Documents
Axygen® deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C. Ultra-flat surface ensures proper sealing with heat sealing film. Clear, V Shaped Well Bottom.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89080-534
  • 89080-532
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Working Well Volume
    Sterility
    Availability
    Price
  • Deep Well Plates
    10 ml
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $122.40
    $122.40
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $612.00
    $612.00
    Prix promotionnel pour les nouveaux clients
    Pack of 5
       $122.40
    Case of 25
       $612.00
    Supplier Number: P-DW-10ML-24-C-S
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89080-534
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Deep Well Plates
    10 ml
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $102.00
    $102.00
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $510.00
    $510.00
    Prix promotionnel pour les nouveaux clients
    Pack of 5
       $102.00
    Case of 25
       $510.00
    Supplier Number: P-DW-10ML-24-C
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89080-532
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Reynosa
    Product available on GSA Advantage®
      Low Manufacturing Impact