ContactUsLinkComponent
Tungsten Lamp for Lambda Spectrometers, PerkinElmer
Catalog # CA97012-474
Supplier:  PerkinElmer
undefined
Tungsten Lamp for Lambda Spectrometers, PerkinElmer
Catalog # CA97012-474
Supplier:  PerkinElmer
Supplier Number:  B0114620
Specifications
  • Description:
    Tungsten Lamp
  • Cat. no.:
    CA97012-474
  • Supplier No.:
    B0114620
Specifications
About this item
PerkinElmer’s long-lifetime deuterium and tungsten source lamps provide wavelength coverage across the working range of the spectrometer for consistently high performance.
  • Long lifetime
  • Broad range of wavelength coverage
  • Consistently high performance
  • Pre-aligned for easy replacement
All lamps are pre-aligned and can be easily changed by the user. Eye protection required when viewing deuterium or tungsten lamps when lit.
Ordering information: Adapter (P/N B0166354) may be required for older models of Lambda 5, 7 and 9.