SP Bel-Art Fluo-Kem® Teflon® FEP Coated Aluminum Scoops, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Aluminum scoop has anti-stick, corrosion-resistant brown Fluo-Kem® Teflon® coating containing FEP, which complies with FDA regulations for food contact.
- Corrosion-resistant
- Anti-stick FEP coating
- Non-slip handle
Handle is not coated so your grip is non-slip.
Frequently Bought Together