Mag-Bind SeqDTR, Omega Bio-Tek
À propos de cet article
Omega Bio-teks Mag-Bind SeqDTR is designed to effective and reliable removal of unincorporated terminators from sequencing reaction. The system combines Omega Bio-teks proprietary chemistry with the reversible nucleic acid-binding properties of paramagnetic beads to eliminate excess nucleotides, primers, salts, and unicorporated dye terminators.
- Long Phred 20 read lengths averaging over 800 bps
- Works with automated liquid handlers
- Can be used in 96 or 384 format
This kit is designed for both manual and fully automated purification of sequencing products, The chemistry has been successfully automated on liquid handlers such has Beckman Coulter’s Biomek FX/NX, and Hamilton STAR/Starlet. Mag-Bind SEQ DTR high recovery rate allows for decreased usage of ABI Big Dye chemistry, normally 1/16 or 1/32 reactions can be performed without a decrease in signal strength when compared to sephadex based clean-up
3 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
Omega Bio-teks Mag-Bind SeqDTR is designed to effective and reliable removal of unincorporated terminators from sequencing reaction. The system combines Omega Bio-teks proprietary chemistry with the reversible nucleic acid-binding properties of paramagnetic beads to eliminate excess nucleotides, primers, salts, and unicorporated dye terminators.
- Long Phred 20 read lengths averaging over 800 bps
- Works with automated liquid handlers
- Can be used in 96 or 384 format
This kit is designed for both manual and fully automated purification of sequencing products, The chemistry has been successfully automated on liquid handlers such has Beckman Coulter’s Biomek FX/NX, and Hamilton STAR/Starlet. Mag-Bind SEQ DTR high recovery rate allows for decreased usage of ABI Big Dye chemistry, normally 1/16 or 1/32 reactions can be performed without a decrease in signal strength when compared to sephadex based clean-up
Specifications for Product Family
Spécifications
Spécifications
- Catalog No:
- CA95059-684
- CA95059-686
- CA95059-688
- Description:
- Mag-Bind SeqDTR
- Mag-Bind SeqDTR
- Mag-Bind SeqDTR
- Target molecule:
- DNA
- DNA
- DNA
- Refine target molecule:
- Amplified/modified DNA
- Amplified/modified DNA
- Amplified/modified DNA
- Sample type:
- PCR and other enzymatic reactions
- PCR and other enzymatic reactions
- PCR and other enzymatic reactions
- Method / Format:
- Magnetic beads
- Magnetic beads
- Magnetic beads
- Prep size:
- Scalable
- Scalable
- Scalable
Spécifications
Product Family Options
Informations sur le produit
- DésignationTailleAvailabilityPrice
- Supplier Number: M1300-05L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.CA95059-684DescriptionMag-Bind SeqDTRTarget moleculeDNARefine target moleculeAmplified/modified DNASample typePCR and other enzymatic reactionsMethod / FormatMagnetic beadsPrep sizeScalable - Supplier Number: M1300-08L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.CA95059-686DescriptionMag-Bind SeqDTRTarget moleculeDNARefine target moleculeAmplified/modified DNASample typePCR and other enzymatic reactionsMethod / FormatMagnetic beadsPrep sizeScalable - Supplier Number: M1300-50L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.Spécifications:
More
Specifications
Cat. No.CA95059-688DescriptionMag-Bind SeqDTRTarget moleculeDNARefine target moleculeAmplified/modified DNASample typePCR and other enzymatic reactionsMethod / FormatMagnetic beadsPrep sizeScalable