ContactUsLinkComponent
SP Bel-Art Disposal Carton for Glass, Bel-Art Products, a part of SP
Catalog # 10027-734
undefined
SP Bel-Art Disposal Carton for Glass, Bel-Art Products, a part of SP
Catalog # 10027-734
Supplier Number:  F24653-0002
Specifications
  • Dimensions:
    20 x 20 x 25 cm (8 x 8 x 10")
  • Cat. no.:
    10027-734
  • Supplier No.:
    F24653-0002
Specifications
About this item
Sturdy corrugated cardboard receptacle is supplied with a 2 mil thick, clear polyethylene bag to contain broken glass fragments and spilled liquids.
  • Corrugated cardboard construction
  • Sturdy design
  • Provided with clear polyethylene containment bag
  • Printed with eye-catching graphics
When the box is full, lift the flap opening to a closed position and the entire unit is ready for disposal. Printed with eye-catching graphics in four languages (English, German, French, Spanish), it is packaged flat to save space and shipping costs and is quickly assembled. Perfect for the benchtop. Does not qualify as a DOT container.