CDC741T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Fournisseur: Hach
Certificats
À propos de cet article
The Radiometer CDC741T is a 2-pole conductivity cell with platinized, platinum sensor (k = 1.0 cm-1), glass body and integrated temperature sensor
The CDC741T sensor's 2 platinum plate design is ideal for use with a sample changer due to easy rinsing. The cell has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters. Please reference article no. E61M023 for a version of the CDC741T with RJ45 plug, compatible for use with Hach sensION+ benchtop conductivity meters.
Frequently Bought Together