CDC511T Conductivity Cell with Temperature Sensor, Radiometer Analytical, Hach
Fournisseur: Hach
Certificats
À propos de cet article
The Radiometer CDC511T is a 4-pole conductivity cell with non-platinized, platinum sensor (k = 1.0 cm-1), integrated temperature sensor and TPX body
The CDC511T sensor's 4 platinum ring design ensures results in general aqueous and non-aqueous media over several decades of conductivity with a single cell, using just one calibration. The CDC511T can be used with a flow-through cell for pure water measurements and meets requirements of USP 24-NF19. It has a fixed cable with MAB6 plug for use with Radiometer Analytical CDM210/230/92 conductivity meters.
Ordering information: Includes Radiometer Analytical CDC511T conductivity cell with Certificate of Conformity.
Frequently Bought Together