SP Bel-Art Stainless Steel Lab Spoon and Spatula, Bel-Art Products, a part of SP
57952-250
- Description:Double-ended, stainless steel, lab spoon/spatula
- Cat. no.:57952-250
- Supplier No.:367110012
This large stainless steel lab spoon is 30.5 cm (12") long for reaching into larger bottles.
- Stainless steel construction
- Long handle provides extra reach
- Two ends are spatula and spoon
One end is a flat spatula measuring 3.1 cm (1¼") wide; the spoon end is 3.7 cm (1½") wide and holds 5 ml. The connecting bar is 14 mm (½") wide.