VWR® Micro Homogenizing Packages
À propos de cet article
The VWR® Standard Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes.
- Designed to mix all sample types
- Quick and efficient processing
Volumes as small as 0.03 mL can be easily processed in a matter of seconds
The VWR® Deluxe Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes. The 5mm flat-bottom generator probe is designed for liquid and soft samples. A 7mm saw-tooth generator probe provides extra cutting action for all samples. The Deluxe Package offers full homogenizing capability for all types of samples within micro-tubes. From light mixing to processing tough tissue samples, this micro-homogenizing package can provide fast and complete breakdown results. Volumes as small as 0.03 mL can be easily processed in a matter of seconds.
The VWR® Premium Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes. The 5mm flat-bottom generator probe is designed for liquid and soft samples. A 7mm saw-tooth generator probe provides extra cutting action for all samples. Complete with stand assembly for hands-free homogenizing of all sample types within micro-tubes. From light mixing to processing tough tissue samples, the Premium micro-homogenizing package can provide fast and complete results without having to hold the homogenizer unit. Volumes as small as 0.03 mL can be easily processed in a matter of seconds.
The 5mm flat-bottom generator probe is designed for liquid and soft samples. Ideal for processing in micro-tubes, whether gently mixing, extracting RNA/DNA, or destroying the cell wall, this standard micro-homogenizing package can effectively and efficiently process virtually any small sample.
Certifications: CE marked.
Ordering information: Supplied with a two-year manufacturer's warranty. All VWR Homogenizing Packages include a carrying case with a foam insert to for protected storage. The Standard Micro-Homogenizing Package (10032-324) includes VWR® 200 Homogenizer unit and 5x75mm flat-bottom generator probe. The Deluxe Micro-Homogenizing Package (10032-326) includes VWR® 200 Homogenizer unit, 5x75mm flat-bottom generator probe, and 7x95mm saw-tooth generator probe. The Premium Micro-Homogenizing Package (10032-328) includes VWR® 200 Homogenizer unit, 5x75mm flat-bottom generator probe, 7x95mm saw-tooth generator probe, and stand assembly.
3 Options Available Below
Product Details & Documents
The VWR® Standard Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes.
- Designed to mix all sample types
- Quick and efficient processing
Volumes as small as 0.03 mL can be easily processed in a matter of seconds
The VWR® Deluxe Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes. The 5mm flat-bottom generator probe is designed for liquid and soft samples. A 7mm saw-tooth generator probe provides extra cutting action for all samples. The Deluxe Package offers full homogenizing capability for all types of samples within micro-tubes. From light mixing to processing tough tissue samples, this micro-homogenizing package can provide fast and complete breakdown results. Volumes as small as 0.03 mL can be easily processed in a matter of seconds.
The VWR® Premium Micro-Homogenizing Package fits into 500µL, 1.5 mL, and 2 mL micro-tubes. The 5mm flat-bottom generator probe is designed for liquid and soft samples. A 7mm saw-tooth generator probe provides extra cutting action for all samples. Complete with stand assembly for hands-free homogenizing of all sample types within micro-tubes. From light mixing to processing tough tissue samples, the Premium micro-homogenizing package can provide fast and complete results without having to hold the homogenizer unit. Volumes as small as 0.03 mL can be easily processed in a matter of seconds.
The 5mm flat-bottom generator probe is designed for liquid and soft samples. Ideal for processing in micro-tubes, whether gently mixing, extracting RNA/DNA, or destroying the cell wall, this standard micro-homogenizing package can effectively and efficiently process virtually any small sample.
Certifications: CE marked.
Ordering information: Supplied with a two-year manufacturer's warranty. All VWR Homogenizing Packages include a carrying case with a foam insert to for protected storage. The Standard Micro-Homogenizing Package (10032-324) includes VWR® 200 Homogenizer unit and 5x75mm flat-bottom generator probe. The Deluxe Micro-Homogenizing Package (10032-326) includes VWR® 200 Homogenizer unit, 5x75mm flat-bottom generator probe, and 7x95mm saw-tooth generator probe. The Premium Micro-Homogenizing Package (10032-328) includes VWR® 200 Homogenizer unit, 5x75mm flat-bottom generator probe, 7x95mm saw-tooth generator probe, and stand assembly.
Specifications for Product Family
Spécifications
- Catalog No:
- 10032-324
- 10032-326
- 10032-328
- Weight:
- 0.64 kg (1.4 lbs.)
- 0.64 kg (1.4 lbs.)
- 0.64 kg (1.4 lbs.)
- Dimensions:
- 5.5 x 19.8 x 5 cm (2.15 x 7.8 x 1.98")
- 5.5 x 19.8 x 5 cm (2.15 x 7.8 x 1.98")
- 5.5 x 19.8 x 5 cm (2.15 x 7.8 x 1.98")
- Power rating:
- 144 W
- 144 W
- 144 W
- Sound level:
- 68 dB
- 68 dB
- 68 dB
Spécifications
Product Family Options
Informations sur le produit
- DésignationElectricalVolume RangeSpeed RangeAvailabilityPrice
- VWR® Standard Micro-Homogenizing Package120 V0.5 - 2 ml tubesAnalog: 5000 - 35000 rpm (with separate on/off switch)Log in pour la disponibilitéKit of 1$3,209.82Spécifications:
More
Specifications
Cat. No.10032-324Weight0.64 kg (1.4 lbs.)Dimensions5.5 x 19.8 x 5 cm (2.15 x 7.8 x 1.98")Power rating144 WSound level68 dB - VWR® Deluxe Micro-Homogenizing Package120 V0.5 - 2 ml tubesAnalog: 5000 - 35000 rpm (with separate on/off switch)KIT(1items)$5,674.74Kit of 1$4,500.36Spécifications:
More
Specifications
Cat. No.10032-326Weight0.64 kg (1.4 lbs.)Dimensions5.5 x 19.8 x 5 cm (2.15 x 7.8 x 1.98")Power rating144 WSound level68 dB - VWR® Premium Micro-Homogenizing Package120 V0.5 - 2 ml tubesAnalog: 5000 - 35000 rpm (with separate on/off switch)Log in pour la disponibilitéKit of 1$4,976.36Spécifications:
More
Specifications
Cat. No.10032-328Weight0.64 kg (1.4 lbs.)Dimensions5.5 x 19.8 x 5 cm (2.15 x 7.8 x 1.98")Power rating144 WSound level68 dB