Phosphate Test Strips, MQuant® | Avantor
Phosphate Test Strips, MQuant®
Phosphate Test Strips, MQuant®
Supplier:  MilliporeSigma
Certificates
Phosphate Test Strips, MQuant®
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
MQuant™ test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: Just dip into sample, add reagent, wait and measure visually against colour scale
  • Immediate results within 2 minutes enables fast decisions
  • Portable: Can be used inside or outside the laboratory
The phosphate test is suitable for determining orthophosphate in wastewater, soil samples, fertilizers and food.
Gradation: 10 - 25 - 50 - 100 - 250 - 500 mg/l PO₄³⁻
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Range
    Type de conditionnement
    No. of Tests
    Availability
    Price
  • Phophate test strips
    1 pc
    10 - 500 mg/L PO₄³⁻
    Plastic Box(es)
    100
    Discontinued
    Supplier Number: 1104280001
  • Phophate test strips
    1 pc
    10 - 500 mg/L PO₄³⁻
    Plastic Box(es)
    100
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $463.85
    $463.85
    Log in pour la disponibilité
    Case of 40
    Case of 40 - $18,554.00
    $18,554.00
    Each
       $463.85
    Case of 40
       $18,554.00
    Supplier Number: 1.10428.0001