Calcium Test Strips, MQuant®
À propos de cet article
MQuant™ test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: Just dip, wait and measure visually against colour scale
- Safe: No auxiliary reagents necessary
- Immediate results within 2 minutes enables fast decisions
- Portable: Can be used inside or outside the laboratory
The calcium concentration is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a colour scale.
Gradation: 10 - 25 - 50 - 100 mg/l Ca
1 Options Available Below
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
MQuant™ test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Easy to handle: Just dip, wait and measure visually against colour scale
- Safe: No auxiliary reagents necessary
- Immediate results within 2 minutes enables fast decisions
- Portable: Can be used inside or outside the laboratory
The calcium concentration is measured semiquantitatively by visual comparison of the reaction zone of the test strip with the fields of a colour scale.
Gradation: 10 - 25 - 50 - 100 mg/l Ca
Product Family Options
Informations sur le produit
- DésignationTailleMéthode chimiqueRangeType de conditionnementNo. of TestsAvailabilityPrice
- Calcium test Strips1 pcGlyoxal bishydroxyanil10 - 100 mg/L CaPlastic Box(es)60Discontinued-Supplier Number: 1100830001L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Calcium test Strips1 pcGlyoxal bishydroxyanil10 - 100 mg/L CaPlastic Box(es)60Each253,76 $Case of 4010 150,40 $Supplier Number: 1.10083.0001L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.