ContactUsLinkComponent
Advanced Purpose Heating Circulators, Grant Instruments
Advanced Purpose Heating Circulators, Grant Instruments
Supplier:  GRANT USA INC.
Certificates
undefined
Advanced Purpose Heating Circulators, Grant Instruments
Supplier:  GRANT USA INC.
Specifications
  • Catalog No:
  • 10029-482
  • 10029-480
  • 10029-484
  • 10029-494
  • 10029-492
  • 10029-496
  • 89260-850
  • 10029-478
  • 89260-844
  • 10029-498
  • 10029-476
  • Heating power:
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • 1.4 kW
  • Temperature resolution:
  • 0.1 °C (0.01 °C with Labwise)
  • 0.1 °C (0.01 °C with Labwise)
  • 0.1 °C (0.01 °C with Labwise)
  • 0.1 °C (0.01 °C with Labwise)
  • 0.1 °C (0.01 °C with Labwise)
  • Temperature stability:
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • ±0.01°C
  • Temperature uniformity:
  • ±0.05°C
  • ±0.05°C
  • ±0.05°C
  • ±0.05°C
  • Timer:
  • 1 min to 99 hrs 59 mins
  • Electrical:
  • 120V, 1.5 kW
  • 120V
  • 120V
  • 120V
  • 120V, 1.5 kW
  • 120V, 1.5 kW
  • 120V, 1.5 kW
  • 120V, 1.5 kW
  • 120V
  • 120V, 1.5 kW
  • 120V, 1.5 kW