Accessories for General Purpose Heating Circulators, Grant Instruments
Supplier: GRANT USA INC.
Accessories for General Purpose Heating Circulators, Grant Instruments
Supplier: GRANT USA INC.
Certificates
À propos de cet article
25 Options Available Below
Product Details & Documents
Specifications for Product Family
Spécifications
- Catalog No:
- 89260-856
- 89260-858
- 89260-860
- 89260-916
- 89260-862
- 89260-864
- 47727-848
- 89260-872
- 89260-868
- 89260-870
- 89260-866
- 66238-596
- 66238-594
- 47728-756
- 97025-578
- 97025-584
- 47729-350
- 89498-146
- 97025-616
- 89498-148
- 89498-138
- 97025-606
- 89498-140
- 97025-608
- 89498-142
- 97025-610
- 89498-144
- 97025-612
- 89260-878
- 89260-880
- 89260-882
- 89260-884
- 89260-894
- 89260-896
- 89260-898
- 89260-900
- 10121-556
- 10121-558
- 10121-560
- Model:
- IC-200 Immersion Cooler
- IC-400 Immersion Cooler
- Cooling power:
- 300W at 20°C, 100W at 40°C
- 350 W at 20 °C, 250 W at 40 °C
- Temperature:
- 0...40 °C
- –20...40 °C
- Min. temperature:
- 0 °C
- –15 °C
- Power consumption:
- 300 W
- 500 W
- Electrical:
- 115V, 50/60 Hz
- 115 V, 50/60 Hz
- Dimensions:
- Coil: 7.6 dia. x 5.7 cm (3 x 2.25")
- Coil: 7.6 dia. x 10.8 cm (3 x 4.25")
Spécifications
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: ST5Spécifications:CSA Certified
More
Specifications
Cat. No.89260-856ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensionsCSA Certified - Supplier Number: ST12Spécifications:CSA Certified
More
Specifications
Cat. No.89260-858ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensionsCSA Certified - Supplier Number: ST18Spécifications:
More
Specifications
Cat. No.89260-860ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - 20 L Refrigeration Unit with Drain TapDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89260-916ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: ST26Spécifications:
More
Specifications
Cat. No.89260-862ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: ST38Spécifications:
More
Specifications
Cat. No.89260-864ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Flat Lid for 5 L Plastic TankDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.47727-848ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: STL38Spécifications:
More
Specifications
Cat. No.89260-872ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: STL12Spécifications:
More
Specifications
Cat. No.89260-868ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: STL26Spécifications:
More
Specifications
Cat. No.89260-870ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: STL5Spécifications:
More
Specifications
Cat. No.89260-866ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: CW5Spécifications:
More
Specifications
Cat. No.66238-596ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Immersion Cooler, 350W, flexible pipe length: 92.7 cm (36.5")Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.66238-594ModelIC-200 Immersion CoolerCooling power300W at 20°C, 100W at 40°CTemperature0...40 °CMin. temperature0 °CPower consumption300 WElectrical115V, 50/60 HzDimensionsCoil: 7.6 dia. x 5.7 cm (3 x 2.25") - Immersion Cooler, 400W, flexible pipe length: 92.7 cm (36.5")Log in pour la disponibilitéEach5 390,96 $Supplier Number: C2GR USSpécifications:
More
Specifications
Cat. No.47728-756ModelIC-400 Immersion CoolerCooling power350 W at 20 °C, 250 W at 40 °CTemperature–20...40 °CMin. temperature–15 °CPower consumption500 WElectrical115 V, 50/60 HzDimensionsCoil: 7.6 dia. x 10.8 cm (3 x 4.25") - Supplier Number: RS14Spécifications:
More
Specifications
Cat. No.97025-578ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: RS28Spécifications:
More
Specifications
Cat. No.97025-584ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: RS38Spécifications:
More
Specifications
Cat. No.47729-350ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: QR-SESpécifications:
More
Specifications
Cat. No.89498-146ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 0.5 ml Tubes (102 Places) not for 5 L TankLog in pour la disponibilitéEach364,98 $Supplier Number: VR-SESpécifications:
More
Specifications
Cat. No.97025-616ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 1.5 ml Tubes (35 Places) for 5 L TankDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89498-148ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 10-13 mm Tubes (30 Places) for 5 L TankDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89498-138ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 10-13 mm Tubes (65 Places) not for 5 L TankLog in pour la disponibilitéEach364,98 $Supplier Number: VR-13Spécifications:
More
Specifications
Cat. No.97025-606ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 16-19 mm Tubes (16 Places) for 5L TankDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89498-140ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 16-19 mm Tubes (36 Places) not for 5 L TankLog in pour la disponibilitéEach364,98 $Supplier Number: VR-19Spécifications:
More
Specifications
Cat. No.97025-608ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 24 mm Tubes (10 Places) for 5 L TankDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89498-142ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 24 mm Tubes (23 Places) not for 5 L TankLog in pour la disponibilitéEach364,98 $Supplier Number: VR-24Spécifications:
More
Specifications
Cat. No.97025-610ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 30 mm Tubes (5 Places) for 5 L TankDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89498-144ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Test Tube Rack for 30 mm Tubes (14 Places) not for 5 L TankLog in pour la disponibilitéEach364,98 $Supplier Number: VR-30Spécifications:
More
Specifications
Cat. No.97025-612ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: T-BTSSpécifications:
More
Specifications
Cat. No.89260-878ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Replacement bridge-plate, fits T optima on ST12, ST18, ST26 and ST38 tanksLog in pour la disponibilitéEach519,82 $Supplier Number: T-BTLSpécifications:
More
Specifications
Cat. No.89260-880ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Replacement bridge-plate, fits T optima to S5 tanksDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89260-882ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Replacement bridge-plate, fits T optima to S12, S18, S26, S38 tanksDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89260-884ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Replacement bridge-plate, fits T opimta to R1, R2 and R3 refrigeration unitsDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89260-894ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Replacement bridge plate, fits T optima to R4 refrigeration unitsDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89260-896ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Replacement bridge-plate, fits T optima onto P5 tanksDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89260-898ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Replacement bridge-plate, fits T optima onto P12 and P18 tanksDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.89260-900ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: LST12Spécifications:
More
Specifications
Cat. No.10121-556ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: LST26Spécifications:
More
Specifications
Cat. No.10121-558ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions - Supplier Number: LST38Spécifications:
More
Specifications
Cat. No.10121-560ModelCooling powerTemperatureMin. temperaturePower consumptionElectricalDimensions
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Clamp to attach Thermostat to Independent Tank; allows independent use of Optima™ Immersion Thermostat, models T100, TC120, TX150, and TXF200Clamp to attach Thermostat to Independent Tank; allows independent use of Optima™ Immersion Thermostat, models T100, TC120, TX150, and TXF20089260-854Log in pour la disponibilitéEach - $268.61
Informations sur le produit
- Catalog #AvailabilityPrice
- Bath Draining Siphon
Informations sur le produit
- Catalog #AvailabilityPrice
- Spheres, Polypropylene (300)