Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Supplier:   Corning
Corning® Single-Use Bags for HYPERStack® Vessels, Corning
Supplier:  Corning
Certificates
Corning® Single-Use Bags for HYPERStack® Vessels, Corning
3 Options Available Below
Product Details & Documents
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Matière
    Availability
    Price
  • Trypsin bag
    5 L
    ULDPE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5l)
    Each (5l) - $412.00
    $412.00
    Each (5l)
       $412.00
    Supplier Number: 91-200-75
  • Quench bag
    5 L
    ULDPE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5l)
    Each (5l) - $438.00
    $438.00
    Each (5l)
       $438.00
    Supplier Number: 91-200-76
  • Media bag
    20 L
    ULDPE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (20l)
    Each (20l) - $400.00
    $400.00
    Each (20l)
       $400.00
    Supplier Number: 91-200-77