SP Bel-Art Large Pails, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Available in yellow polyethylene or autoclavable translucent polypropylene, these 13.2 liter (14qt) pails have a snug-fitting lid and plated steel bail.
- Yellow polyethylene or translucent polypropylene construction
- Polypropylene pail is autoclavable
- Snug-fitting lid
- Plated steel bail
Each bucket is 32 cm (12¾") high with a 27 cm (10½") I.D. at top. Pail and cover 70690-026 are steam autoclavable at 121ºC (250ºF).
Certifications: Meets US Specification MIL-P36257A.
Frequently Bought Together