SP Bel-Art McAlpine™ ABS Sink 'S' and 'P' Trap
ABS drain traps withstand frost, boiling water, detergents and chemicals and include PVC plug, strainer, waste body, waste fitting, back nut, sealing cup and compression for copper tubing.
Can be configured as an 'S' or 'P' trap
ABS plastic construction
- Waste section features PVC plug, washer, and nut
- Easy installation
Waste section at top consists of a threaded body with PVC plug, washer and nut. Requires no special tools for installation or maintenance. Available in 3.8 cm (1½") size only.
Frequently Bought Together