EASYstrainer Cell Sieves, Greiner Bio-One
EASYstrainer Cell Sieves, Greiner Bio-One
EASYstrainer Cell Sieves, Greiner Bio-One
EASYstrainer Cell Sieves, Greiner Bio-One
EASYstrainer cell sieves are filters for cell suspensions and primary cell isolates.
  • Range of cell sizes for cells and tissues
  • Flexible fit suitable for tubes 1.5 ml to 50 ml
  • Invertible upper part of EASYstrainer Small for recovery of retained cells
  • Handle and surrounding rim fascilitates aseptic working and reduces risk of contamination
  • Ventilation Gap allows for fast filtration and prevents overflow
  • Stackable design allows for sequential filtration from large to small mesh sizes
  • single packaging fascilitates aseptic working and reduces risk of contamination
 : Individual sterile blister packs.
Product Family Options
  • Désignation
    Sterility
    Color
  • EASYstrainer for 50 ml tubes, 40 μm mesh size
    Sterile
    Green
    Case of 50
    Case of 50 - $209.09
    $209.09
    Case of 50
       $209.09
     : 542040
  • EASYstrainer for 50 ml tubes, 70 μm mesh size
    Sterile
    Blue
    Case of 50
    Case of 50 - $208.78
    $208.78
    Case of 50
       $208.78
     : 542070
  • EASYstrainer for 50 ml tubes, 100 μm mesh size
    Sterile
    Yellow
    Case of 50
    Case of 50 - $209.06
    $209.06
    Case of 50
       $209.06
     : 542000
  • EASYstrainer Small for 1.5 ml to 15 ml tubes, 100 μm mesh size
    Sterile
    Yellow
    Case of 50
    Case of 50 - $315.59
    $315.59
    Case of 50
       $315.59
     : 542100
  • EASYstrainer Small for 1.5 ml to 15 ml tubes, 20 μm mesh size
    Sterile
    Red
    Case of 50
    Case of 50 - $321.76
    $321.76
    Case of 50
       $321.76
     : 542120
  • EASYstrainer Small for 1.5 ml to 15 ml tubes, 40 μm mesh size
    Sterile
    Green
    Case of 50
    Case of 50 - $315.59
    $315.59
    Case of 50
       $315.59
     : 542140
  • EASYstrainer Small for 1.5 ml to 15 ml tubes, 70 μm mesh size
    Sterile
    Blue
    Case of 50
    Case of 50 - $332.93
    $332.93
    Case of 50
       $332.93
     : 542170