ContactUsLinkComponent
Bead Ruptor Pre-Filled Bead Tubes, 30 ml, Omni International
Fournisseur:  OMNI INTERNATIONAL
Certificats
undefined
Bead Ruptor Pre-Filled Bead Tubes, 30 ml, Omni International
Fournisseur:  OMNI INTERNATIONAL
Spécifications
  • Catalog No:
  • 10032-522
  • Processing range:
  • 5 mL to 30 mL
  • Temperature range:
  • –80 to 121°C