ContactUsLinkComponent
750W Ultrasonic Power Supply, Ace Glass Incorporated
  89075-480
 :  Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
undefined
750W Ultrasonic Power Supply, Ace Glass Incorporated
  89075-480
 :  Ace Glass
 :  9810-24
Product available on GSA Advantage®
Small Business Enterprise
  • Description:
    Ultrasonics Vibra-Cell Power Supply, 750 W
  • Cat. no.:
    89075-480
  • Supplier No.:
    9810-24
Ultrasonic power supply, 750W, for superior mixing with automatic amplitude and frequency control circuitry that eliminates the need for constant adjustments, assuring optimum cavitation at any power level
  • Exclusive energy (Joule) setpoint circuit
  • Nonvolatile memory function for storing up to ten preset operating programs
  • Tactile keypad with user friendly menu driven LCD display
  • Elapsed time/run time timer
  • Power (watts) readout
  • Integral temperature controller
New space-saving design with auto tuning that matches the power supply to the converter/probe assembly and does not have to be manually tuned each time the probe is changed or the unit is turned on. Three-year unconditional warranty on power supply and converter. Supplied with detailed operating instructions. not supplied with horn or glass reactors (order separately).