SP Bel-Art Microscope Slide Dispenser, Bel-Art Products, a part of SP
Catalogue # 13197-278
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Dimensions:11.1 x 12.4 x 14 cm (43/8 x 47/8 x 51/2")
- Cat. no.:13197-278
- Supplier No.:F445620000
Spécifications
À propos de cet article
ABS plastic dispenser releases clean, dust-free 25 x 75 mm (1 x 3") slides individually by the turn of a knob on either side.
- Clear plastic cover for easy visual check on slides
- Cover pulls off vertically for refilling
The dispenser holds approximately 72 slides (depending on thickness).