ContactUsLinkComponent
SP Bel-Art Round-Bottom Flask Support, Bel-Art Products, a part of SP
Catalogue # 13196-074
undefined
SP Bel-Art Round-Bottom Flask Support, Bel-Art Products, a part of SP
Catalogue # 13196-074
Supplier Number:  F38950-0000
Spécifications
  • Ø:
    17.1 cm (6 3/4")
  • Height:
    5.1 cm (2")
  • Cat. no.:
    13196-074
  • Supplier No.:
    389500000
Spécifications
À propos de cet article
Round-bottom flasks of any size up to 10 liters are cradled securely in the stepped concentric rings of this holder that can be stacked together for convenient storage.
  • Round-bottomed design
  • Polypropylene construction
  • Corrosion-resistant
  • Autoclavable
Polypropylene plastic rack is not affected by spilled acids, alkalis or other corrosive liquids. Steam autoclavable at 121°C (250°F).