Qualitative Filter Paper, Grade 617, Ahlstrom
Supplier: Ahlstrom
Certificates
About this item
Grade 617 Qualitative Filter Papers are high-purity papers with high-speed filtration capabilities
They are made from 60% cotton and 40% wood fibers.
Specifications
- Catalog No:
- 30617-438
- 30617-440
- 28342-028
- 28342-032
- 75875-630
- 47745-830
- 47745-832
- 97058-876
- 47745-824
- 47745-826
- 47745-822
- 30617-442
- Weight:
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- 123 g/m2
- Thickness:
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- 0.51 mm
- Filtration speed:
- 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg) - 360 mL/min (Rapidity)
19 s/100mL (Herzberg)
- Capillary rise:
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- 48 mm
- Flow rate:
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Fast
- Load capacity:
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Very High
- Retention:
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm
- 35 µm