Halogen Bulbs, Bulbtronics©
Supplier: Bulbtronics
Jump to:
À propos de cet article
Various halogen bulbs.
7 Options Available Below
Product Details & Documents
Various halogen bulbs.
Product Family Options
Informations sur le produit
- DésignationFilamentBulb LifeLongueurØModèleFondTensionPuissanceOEM NumberAvailabilityPrice
- Bulb, Bulbrite, 40 mm Light Center LengthCC8/coiled coil axial2000 h53 mm12.7 mmT4GY6.35/2-pin120 V35 W652035Discontinued-Supplier Number: 0002372
- Bulb, Eiko, 34.8 mm Light Center LengthCC8/coiled coil axial1000 h62 mm12.7 mmT4BA15D/DC Bayonet120 V150 W15246Discontinued-Supplier Number: -
- Bulb, Eiko, 19.56 mm Light Center LengthC6/single coil transverse100 h30 mm9.53 mmT3G4/2-pin6 V20 W10330Discontinued-Supplier Number: -
- Bulb, Eiko, Dichroic ReflectorC6/single coil transverse3000 h47 mm50.8 mmMR16GX5.3/2-pin12 V20 W15070Discontinued-Supplier Number: 0000505
- Bulb, Eiko, Dichroic ReflectorCC6/coiled coil transverse200 h44.45 mm50.8 mmMR16GX5.3/2-pin10.8 V30 W02370Discontinued-Supplier Number: -
- Bulb, Eiko, Faceted Dichroic ReflectorCC6/coiled coil transverse4000 h44.45 mm50.8 mmMR16GX5.3/2-pin12 V42 W15034Discontinued-Supplier Number: -
- Bulb, Eiko, Dichroic ReflectorCC6/coiled coil transverse4000 h50.8 mm50.8 mmMR16GX5.3/2-pin12 V50 W15080Discontinued-Supplier Number: 0001596
- Bulb, Eiko, Dichroic ReflectorCC6/coiled coil transverse500 h44.45 mm50.8 mmMR16GX5.3/2-pin20 V150 W10146Discontinued-Supplier Number: 0001193
- Bulb, Eiko, Dichroic ReflectorCC6/coiled coil transverse40 h47 mm50.8 mmMR16GX5.3/2-pin21 V150 W10264Discontinued-Supplier Number: 0001367
- Bulb, Eiko, Dichroic ReflectorCC6/coiled coil transverse200 h44.45 mm50.8 mmMR16GX5.3/2-pin21 V150 W10275Each$99.04Pack of 10$990.40Supplier Number: 0001405
- Bulb, Eiko, Dichroic ReflectorCC6/coiled coil transverse50 h44.45 mm50.8 mmMR16GX5.3/2-pin25 V250 W02410Discontinued-Supplier Number: -
- Bulb, Eiko, Dichroic ReflectorC6/single coil transverse2000 h35.56 mm34.98 mmMR11GZ4/2-pin12 V12 W15115Discontinued-Supplier Number: -
- Bulb, General Electric, 30 mm Light Center LengthCBAR6/C6F/single coil flat/oval transverse50 h50.8 mm12.7 mmT4G6.35/2-pin24 V150 W13598Discontinued-Supplier Number: -
- Bulb, General ElectricC6/single coil transverse6000 h47.63 mm50.8 mmMR16GU5.3/2-pin12 V50 W20871Discontinued-Supplier Number: -
- Bulb, General ElectricC6/single coil transverse6000 h47.63 mm50.8 mmMR16GX5.3/2-pin12 V50 W20833Discontinued-Supplier Number: -
- Bulb, General Electric, 22.23 mm Light Center LengthC6/single coil transverse4000 h47.63 mm50.8 mmMR16GX5.3/2-pin12 V71 W20843Discontinued-Supplier Number: -
- Bulb, General Electric, Faceted Dichroic ReflectorCC6/coiled coil transverse50 h44.45 mm50.8 mmMR16GX5.3/2-pin24 V250 W37462Discontinued-Supplier Number: -
- Bulb, General Electric, 30 mm Light Center LengthCBAR6/C6F/single coil flat/oval transverse50 h44.45 mm9.53 mmT3GY6.35/2-pin12 V100 W14876Discontinued-Supplier Number: -
- Bulb, General ElectricC8/single coil axial2000 h119 mm7.87 mmT2 1/2R7S/RSC recessed single contact120 V350 W13894Discontinued-Supplier Number: -
- Bulb, Gilway, 30.23 mm Light Center LengthC6/single coil transverse2000 h55.12 mm12.7 mmT4GY9.5/2-pin prefocus12 V20 WL9404Discontinued-Supplier Number: -
- Bulb, Gilway, 30.23 mm Light Center LengthC6/single coil transverse2000 h-12.7 mmT4GY9.5/2-pin prefocus12 V20 WL9404-K2Discontinued-Supplier Number: -
- Bulb, Higuchi, 33 mm Light Center LengthCC2V1500 h45 mm12.7 mmT4E11/mini-candelabra110 V/130 V50 WJD-7003Log in pour la disponibilitéEach$18.65Supplier Number: 0000395
- Bulb, Higuchi, Dichroic ReflectorCC2V2000 h75 mm50.8 mmMR16E17/intermediate screw110 V/120 V50 WJDR-9125Discontinued-Supplier Number: -
- Bulb, HiguchiC6/single coil transverse1500 h35.56 mm35 mmMR11G4/2-pin12 V10 WJCR-8297Each$19.22Case of 10$192.20Supplier Number: 0002301
- Bulb, HiguchiC6/single coil transverse1500 h35.56 mm35 mmMR11G4/2-pin12 V5 WJCR-8295Discontinued-Supplier Number: -
- Bulb, HiguchiC6/single coil transverse2000 h31 mm7.87 mmT2 1/2G4/2-pin12 V20 WJC-5017Discontinued-Supplier Number: -
- Bulb, HiguchiC6/single coil transverse2000 h31 mm9.53 mmT3G4/2-pin12 V10 WJC-5011Each$46.52Case of 10$465.20Supplier Number: 0002109
- Bulb, HiguchiC6/single coil transverse2000 h31 mm9.53 mmT3G4/2-pin6 V10 WJC-5605Log in pour la disponibilitéEach$91.41Supplier Number: 0002138
- Bulb, HiguchiC6/single coil transverse1000 h31 mm9.53 mmT3G4/2-pin24 V20 WJC-5517Log in pour la disponibilitéEach$197.24Supplier Number: 0002131
- Bulb, Higuchi, 20.07 mm Light Center LengthC6/single coil transverse2000 h33 mm11.94 mmT4G4/2-pin6 V20 WJC-5607Discontinued-Supplier Number: -
- Bulb, Higuchi, Dichroic ReflectorCC2V2000 h55.12 mm50.8 mmMR16GU10/twist and lock110 V/120 V50 WJDR-9825FPDiscontinued-Supplier Number: -
- Bulb, HiguchiC6/single coil transverse2000 h44 mm9.53 mmT3GY6.35/2-pin12 V100 WJC-5057Each$9.25Case of 10$92.50Supplier Number: 0000486
- Bulb, HiguchiC6/single coil transverse2000 h44 mm11.94 mmT4GY6.35/2-pin12 V50 WJC-5043Discontinued-Supplier Number: -
- Bulb, HiguchiC8/single coil axial2600 h118 mm9.53 mmT3R7S/RSC recessed single contact120 V500 WJ-2027Discontinued-Supplier Number: -
- Bulb, HiguchiC8/single coil axial2000 h118 mm9.53 mmT3R7S/RSC recessed single contact230 V500 WJ-3119Discontinued-Supplier Number: -
- Bulb, Eiko, Dichroic ReflectorCC6/coiled coil transverse40 h44.45 mm50.8 mmMR16GX5.3/2-pin21 V150 W02290Discontinued-Supplier Number: -